Microwave Reaction Vials, Ace Glass Incorporated
Supplier: Ace Glass
5704-02 5704-05 5704-10
89427-726CS 717.8 USD
89427-726 89427-728 89427-730
Microwave Reaction Vials, Ace Glass Incorporated
Vials
Heavy wall, Type 1 clear borosilicate glass for higher pressure, microwave digestion.
20 mm flange top to accomodate crimp seals. Available with either conical or rounded bottoms.