Anti-Porcine Serum Rabbit Antibody

Supplier: MP Biomedicals
08650351
IC650351EA 169.51 USD
IC650351
Anti-Porcine Serum Rabbit Antibody
Antibodies
Anti-Porcine
Host: rabbit
Form: Lyophilized
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR