E. coli Pyrophosphatase
Supplier: New England Biolabs (NEB)
M0361S M0361L
101641-572EA 73 USD
101641-572 101641-570
E. coli Pyrophosphatase
Enzymes
Inorganic pyrophosphatase (PPase) catalyzes the hydrolysis of inorganic pyrophosphate to form orthophosphate
P207–4 + H20 → 2HP04–2. Pyrophosphatase, Inorganic (E. coli) is prepared from a clone of the E. coli inorganic pyrophosphatase gene.