K. lactis GG799 Competent Cells, New England Biolabs
Supplier: New England Biolabs
C1001S
101227-510EA 463 USD
101227-510
K. lactis GG799 Competent Cells, New England Biolabs
Competent Cells
Chemically competent Kluyveromyces lactis cells suitable for transformation with a linearized pKLAC-series expression vector
GG799 cells are characterized by very high cell density growth and efficient expression of foreign proteins. GG799 cells have no genetic modifications.