Accessories for VACUUBRAND® Rotary Vane Vacuum Pumps, Brand
Supplier: Brand
698017 20698007 20698000 662518 20698006 662532 662511 698003
89178-210EA 1476.2 USD
89178-210 89178-178 89178-202 89178-204 89178-180 89178-206 89178-182 89178-208
Accessories for VACUUBRAND® Rotary Vane Vacuum Pumps, Brand
Pumps Vacuum Pumps
Help protect your vacuum pump and laboratory environment with these accessories for VACUUBRAND® rotary vane and Chemistry-HYBRID vacuum pumps
Inlet Catchpot/Separator AK; inlet catchpots help protect the pump from condensate droplets and particles. Wetted materials are PMP and aluminum.
Oil Mist Filter FO; oil mist filter helps preserve the lab atmosphere by capturing oil mist generated at the outlet of a rotary vane pump and depositing it into a catchpot. These highly efficient filters allow for easy draining of captured oil, and have an integrated pressure relief valve in case of filter blockage. Wetted materials are Aluminum, PMP and fiberglass epoxy.
Hose Flanges; premium quality machined aluminum flanges with hose barbs or nipples for adapting VACUUBRAND® and other vacuum pumps to tubing.
Catchpots and oil mist filters are designed to integrate neatly with VACUUBRAND® for best performance and aesthetics.