Accessories for Extrelut® NT, MilliporeSigma
Supplier: MilliporeSigma
1.15094.0001 1.15095.0001 1.15096.0001 1.14236.0001 1.14237.0001 1.14567.0001
EM1.15094.0001EA 485.96 USD
EM1.15094.0001 EM1.15095.0001 EM1.15096.0001 52428-676 52428-678 52428-680
Accessories for Extrelut® NT, MilliporeSigma
Chromatography Columns
Extrelut® NT simplifies liquid-liquid extraction in its most effective form by using a chemically inert, wide-pore and highly pure diatomaceous earth based solid phase.
Depending on the sample volume three types of columns are available: Extrelut® NT1 can take up a maximum of 1 ml, Extrelut® NT3 3 ml and Extrelut® NT20 up to 20 ml of sample.