Flushing Adapter, Glassco
Supplier: Glassco
048.202.01
89426-436EA 129.83 USD
89426-436
Flushing Adapter, Glassco
Adapters Adapters, Conical Ground Joints
The flushing adapter is used to flush and fill attached vessels.
  • T-bore PTFE stopcock
  • [ST] joint
Adapter is equipped with a 2mm T-bore PTFE stopcock.