Filter Funnel, Buchner, Glassco
Supplier: Glassco
256.C00.00 256.C00.01 256.C00.02A 256.C00.03A 256.C00.03B 256.C00.04A 256.F00.00 256.F00.01 256.F00.02A 256.M00.00 256.M00.00A 256.M00.01 256.M00.02A 256.M00.03A 256.M00.03B
89426-708EA 46.74 USD
89426-708 89426-710 89426-712 89426-714 89426-716 89426-718 89426-720 89426-722 89426-724 89426-726 89426-728 89426-730 89426-732 89426-734 89426-736
Filter Funnel, Buchner, Glassco
Funnels Filtration Funnels Büchner Funnels
Funnels feature a sealed-in glass fritted disc of designated porosity
Useful wherever filter paper or membranes cannot withstand the chemical attack of the filtrate.