Chromolith® Cartridges, MilliporeSigma
Supplier: MilliporeSigma
Chromolith®
1.52009.0001 1.52035.0001 1.52008.0001 1.52004.0001 1.52008.0001 1.52005.0001 1.52036.0001 1.52011.0001 1.52011.0001 1.52036.0001 1.25262.0001 1.52037.0001 1.52037.0001 1.52013.0001 1.25262.0001 1.52030.0001 1.52030.0001 1.52005.0001 1.52004.0001 1.52013.0001 1.52035.0001
EM1.52011.0001EA 653.34 USD
EM1.52011.0001 CA1.52036.0001 CA1.52030.0001 EM1.52036.0001 CA97025-828 CA1.52035.0001 EM1.52013.0001 CA1.52011.0001 EM1.52037.0001 CA1.25262.0001 97025-824 10143-314 CA97025-822 CA1.52037.0001 97025-822 EM1.52030.0001 CA97025-826 EM1.52035.0001 97025-826 CA97025-824 CA1.52013.0001
Chromolith® Cartridges, MilliporeSigma
Chromatography Columns
Although the Chromolith® columns are well known for their robustness and longevity, guard columns are available to prolong the lifespan of Chromolith® analytical columns even further.
The guard cartridge kits includes the guard cartridge holder and three guard cartridges.
Ordering information: Chromolith® RP-18 endcapped 5-2 guard cartridge kit includes 3 guard cartridges and 1 guard cartridge holder.