VWR® Plain Apron Tables
Supplier: VWR International
89375-150 89375-152 89375-154 89375-156 89375-158 89375-160 89375-162 89375-164 89375-166 89375-168 89375-170 89375-172 89375-238 89375-242 89375-244 89375-246 89375-252 89375-254 89375-260 89375-264 89375-266 89375-270 89375-272 89375-276 89375-310 89375-312 89375-314 89375-316 89375-318 89375-320 89375-328 89375-330 89375-332 89375-374 89375-378 89375-380 89375-382 89375-388 89375-390 89375-396 89375-400 89375-408 89375-412
89375-150EA 1169.74 USD
89375-150 89375-152 89375-154 89375-156 89375-158 89375-160 89375-162 89375-164 89375-166 89375-168 89375-170 89375-172 89375-238 89375-242 89375-244 89375-246 89375-252 89375-254 89375-260 89375-264 89375-266 89375-270 89375-272 89375-276 89375-310 89375-312 89375-314 89375-316 89375-318 89375-320 89375-328 89375-330 89375-332 89375-374 89375-378 89375-380 89375-382 89375-388 89375-390 89375-396 89375-400 89375-408 89375-412
VWR® Plain Apron Tables
Furniture Tables and Workbenches
Plain apron tables are available in a variety of sizes, with several different top materials.
  • Available in two heights: 30 or 36"
  • Finished with a chemical resistant, earth-friendly UV finish
  • Legs and aprons are constructed of solid red oak, and are smoothly machined
  • Black rubber leg boots which offer protection against scuffing and chemical spills
  • Equipped with non-skid adjustable glides for precise leveling and for prevention of rocking on uneven floors
Aprons are constructed of solid red oak with heavy-duty 13-gauge steel corner braces. 2¼" squareMore Product Information