Kimtech™ G3 EvT Nitrile Gloves, Kimberly-Clark Professional
Supplier: Kimberly-Clark
38702 38705
89423-030CS 3146.2 USD
89423-030 89423-032 89423-034 89423-036 89423-038 89423-040
Kimtech™ G3 EvT Nitrile Gloves, Kimberly-Clark Professional
Gloves
In the cleanroom environment, one small particle can mean big trouble. One spec of dust on a worker’s glove can cause costly damage, therefore it's critical to have a glove with low particles and extractables, such as the Kimtech™ G3 EvT nitrile gloves.
  • Beaded cuffs for easy donning
  • Low particles and extractables
  • Not made with silicone or natural rubber latex
Kimtech™ G3 EvT nitrile gloves are designed with 'Extra-Value Technology' to have low particles and extractables. They are cleaned to meet the needs of a critical cleanroom environment. Durable and powder-free.
Gloves have excellent overall tactile performance. They are not made with silicone or natural rubber latex. Additionally, the textured and tacky surface provides increased dexterity for critical tasks in semiconductor, electronics, and medical device manufacturing facilities. They have an AQL of 1.5 for pinholes and can even be recycled.
Certifications: Exceeds the ASTM D 5151 standard for critical defects or pinholes.Recommended for ISO Class 3 and higher environments.Meets U.S. FDA regulations for food-handling applications.