Peptide Vessel with Threaded Top and PTFE Stopcock, Ace Glass Incorporated
Supplier: Ace Glass
6402-12 6402-15 6402-20 6402-24 6402-27
89054-182EA 290.76 USD
89054-182 89054-184 89054-186 89054-188 89054-190
Peptide Vessel with Threaded Top and PTFE Stopcock, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Cylindrical Shape
Cylindrical vessel is equipped with a 2mm bore PTFE stopcock and porosity C (25–50µm) fritted disc at bottom, and a GPI thread at top
Supplied with solid cap, holed cap, and PTFE-faced silicone rubber septum. All wetted surfaces are either glass or PTFE.