Rotary Evaporator Traps, Ace Glass Incorporated
Supplier: Ace Glass
3971-21 3971-01 3971-05 3971-03
89214-956EA 1160.62 USD
89214-956 89215-164 89215-166 89215-168
Rotary Evaporator Traps, Ace Glass Incorporated
Vacuum Traps
Replacement borosilicate glass components for Buchi® R-220, R-220EX, and R-220SE rotary evaporators.
Ordering information: Inner and outer cold trap components are available poly-coated or plain, non-coated. DN40 inner ball joint includes CAPFE (PTFE encapsulated silicone rubber) O-Rings (89061-130), size –225. Black cap (included) is SVL-22 thread. Top CAPFE O-Ring (89061-168 - included) for 89215-168 and 89215-166 is for 150mm grooved top flat flange. The PFA cap must be ordered separately.