Anti-ANGPTL2 Rabbit Polyclonal Antibody

Supplier: AdipoGen Life Sciences
AG-25A-0068-C100
102514-542EA 221.54 USD
102514-542
Anti-ANGPTL2 Rabbit Polyclonal Antibody
Antibodies
ANGPTL2 stimulates expansion of HSCs. ANGPTL2 plays an important role in the pathogenesis of diabetic glomerulopathy. The upregulation of ANGPTL2 in diabetic glomerulopathy shows a close relationship to abnormal microvasculature and endothelial inflammation.

Type:
Antigen: ANGPTL2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR