Anti-FOXP3 Rat Monoclonal Antibody [clone: MF333F]

Supplier: AdipoGen Life Sciences
AG-20A-0025-C050 AG-20A-0025-C100
102514-006EA 332.31 USD
102514-006 102514-004
Anti-FOXP3 Rat Monoclonal Antibody [clone: MF333F]
Antibodies
FOXP3 is involved in immune system responses. It functions as the master regulator in the development and function of regulatory T cells. FOX proteins belong to the forkhead/winged-helix family of transcriptional regulators and are presumed to exert control via similar DNA binding interactions during transcription. Defects in FOXP3 are the cause of immunodeficiency polyendocrinopathy, enteropathy, X-linked syndrome (IPEX); also known as X-linked autoimmunity-immunodeficiency syndrome. IPEX is characterized by neonatal onset insulin-dependent diabetes mellitus, infections, secretory diarrhea, trombocytopenia, anemia and eczema.

Type:
Antigen: FOXP3
Clonality: Monoclonal
Clone: MF333F
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR