IR Thermometer Guns, 8:1, Sper Scientific
Supplier: Sper Scientific
800101 800102
97035-832EA 204.38 USD
97035-832 97035-830
IR Thermometer Guns, 8:1, Sper Scientific
Thermometers Digital Thermometers
Rugged and ergonomically designed these infrared thermometer guns come complete with batteries and measures up to 605°F (320°C) or 930°F (500°C).
  • Results are automatically held for 6 seconds after the trigger is released
  • Has auto power off
  • 8:1 distance to spot ratio (D:S)
Simply hold the IR gun, press the trigger and aim the laser guide at the target. Results immediately appear in ºC or ºF on the backlit LCD.
IR thermometers are used to safely measure the temperature in: Inaccessible, hot or hazardous objects in electrical, hazmat, HVAC, and other fields; moving machinery in factories, automobiles and elsewhere; electrical troubleshooting HVAC energy audits; avoiding contamination in the food, beverage and pharmaceutical industries.