H-Trp-βNA

Supplier: BACHEM AMERICAS INC

4002361.0001 4002361.0005
K-1525.0001BAEA 184.23 USD
K-1525.0001BA K-1525.0005BA
H-Trp-βNA
H-Trp-βNA

Sequence: H-Trp-βNA

Formula: C₂₁H₁₉N₃O
Storage Temperature: Ambient
MDL Number: MFCD00051194
CAS Number: 3326-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR