L-Thiolhistidine

Supplier: BACHEM AMERICAS INC

4014107.0001 4014107.0005
F-3615.0001BAEA 1049 USD
F-3615.0001BA F-3615.0005BA
L-Thiolhistidine
L-Thiolhistidine

Sequence: H-2-Mercapto-His-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Key intermediate in the synthesis of the naturally occurring amino acid L-(+)-ergothioneine and analogs.

Formula: C₆H₉N₃O₂S
Storage Temperature: Freezer
MDL Number: MFCD00047018
CAS Number: 2002-22-4
EINECS: 217-899-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR