(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid
Supplier: BACHEM AMERICAS INC
Synonyms:
(S)-N-FMOC-Octylglycine, N-Fmoc-2-octyl-L-glycine, Fmoc-octyl-Gly-OH
4062518.0500
4062518.1000
B-4060.0500BAEA
571.91
USD
B-4060.0500BA
B-4060.1000BA
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid
Sequence: Fmoc-octyl-Gly-OH
Bachem offers derivatives of α-aminoglycine, cyclohexylglycine, cyclopentylglycine, and the PNA building block N-β-aminoethylglycine. For t-butylglycine derivatives see t-Leucine.
Synonym(s): Fmoc-(S)-2-aminodecanoic acid#Fmoc-L-decyline#Fmoc-L-2-aminocapric acid
Formula:
C₂₅H₃₁NO₄ MW: 409.526 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01311753 CAS Number: 193885-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...