iTaq™ DNA Polymerase, Bio-Rad
Supplier: BIO-RAD LABORATORIES MS
1708870 1708875
101645-374EA 540 USD
101645-374 101645-380
iTaq™ DNA Polymerase, Bio-Rad
Nucleic Acid Reagents End-point PCR Enzymes and Kits
iTaq™ DNA polymerase is an antibody-mediated hot-start DNA polymerase suitable for both PCR and real-time PCR applications. The enzyme is activated after a 3min denaturation step at 95°C. iTaq DNA polymerase is highly specific, sensitive, and easy to use.
Other benefits include:
* Improved product yields compared to non-antibody-based DNA polymerases
* Amplification of a wide range of targets
* Convenient reaction assembly at room temperature
* Improved product yields compared to non-antibody-based DNA polymerases