Glass Econo-Column Columns, Bio-Rad
Supplier: Bio-Rad
7370517 7371022 7371522 7371552 7374150 7374253 7375031 7374153 7376151 7370512 7374011 7374151 7376131 7371593 7372522 7374006 7376601 7370507 7370707 7371507 7372507 7372593 7374021 7374152 7374252 7375021 7375051 7375071 7376116 7371007 7371532 7371576 7371591 7372512 7372532 7374156 7370752 7371052 7372551 7370732 7371012 7371093 7371517 7372576 7372591 7374251 7376108 7370712 7370717 7371032 7371091 7371512 7374711 7375011 7376607
101646-750EA 129.23 USD
101646-750 101646-770 101646-786 101646-790 101646-826 101646-834 101646-844 101646-852 101646-870 101646-752 101646-822 101646-832 101646-882 101646-748 101646-818 101646-828 101646-848 101646-850 101646-880 101646-796 101646-804 101646-816 101646-886 101646-746 101646-754 101646-780 101646-798 101646-800 101646-814 101646-820 101646-830 101646-842 101646-846 101646-864 101646-868 101646-872 101646-874 101646-878 101646-766 101646-788 101646-792 101646-794 101646-802 101646-806 101646-824 101646-836 101646-764 101646-774 101646-808 101646-838 101646-858 101646-762 101646-768 101646-778 101646-784 101646-810 101646-812 101646-840 101646-860 101646-876 101646-756 101646-758 101646-760 101646-772 101646-776 101646-782 101646-854 101646-856 101646-862 101646-866 101646-888
Glass Econo-Column Columns, Bio-Rad
Chromatography Columns
Econo-Column™ chromatography columns are high-quality, low-pressure, glass chromatography columns. These highly versatile columns range in size from 5 to 170 cm long and 0.5 to 5.0 cm in diameter. A porous polymer bed support at the bottom of the column retains fine particles, and translucent polypropylene end fittings allow viewing of the entire column bed. Econo-Column chromatography columns are packaged in quantities of one, two, and four, depending on column diameter and length. All packages contain one stopcock per column ordered.
Jacketed Econo-Column columns are glass Econo-Columns with an integral water jacket for cooling or heating. Jacketed columns are used in applications requiring temperature control.
Accessories: Jacketed columns are used in applications requiring temperature control.
Features:
* Simple packing and procedure running
* Autoclavability and NaOH compatibility
* Operation at pressures <1 bar (14.7 psi) or in gravity flow applications * Simple packing and procedure running
* Autoclavability and NaOH compatibility
* Operation at pressures <1 bar (14.7 psi) or in gravity flow applications
Ordering information: Reservoirs are available in 500 ml and 1 L capacities and will fit 0.5, 0.7, 1.0, or 1.5 cm Int. Ø Econo-Column chromatography columns. To make a constant-pressure reservoir, close the reservoir top with a stopper that contains a piece of glass tubing extending into the reservoir. The removable upper cap has a male Luer-Lok fitting. See Accessories tab.