Affi-Gel 10 and Affi-Gel 15 Media, Bio-Rad
Supplier: 0000013515
1536052 1536099 1536098 1536051
101642-964EA 1553.85 USD
101642-964 101642-974 101642-972 101642-962
Affi-Gel 10 and Affi-Gel 15 Media, Bio-Rad
Protein Purification Protein Purification Resins
Affi-Gel 10 and Affi-Gel 15 gels are activated affinity media that provide spontaneous, rapid, and highly efficient coupling of ligands via primary amines
  • Aqueous or anhydrous coupling conditions
  • Complete protein coupling within 4 hours at 4 °C
  • Protein coupling capacity up to 35 mg/ml gel
Affi-Gel 10 medium is most efficient for coupling neutral or basic proteins with pI from 6.5 to 11. Affi-Gel 15 medium is recommended for coupling acidic proteins with pI <6.5. Affi-Gel 10 and Affi-Gel 15 media offer: