Potassium peroxodisulfate, powder pillow

Danger

Synonyms: Dipotassium peroxodisulphateDipotassium peroxodisulfatePeroxydisulphuric acid dipotassium saltPotassium persulfatedi-Potassium peroxodisulfatePeroxydisulfuric acid dipotassium saltPotassium persulphatedi-Potassium peroxodisulphate

245199 245199
89230-940PK 91.83 USD
89230-940 CA2451-99
Potassium peroxodisulfate, powder pillow
Potassium peroxodisulphate

For determination of total phosphate by acid persulfate digestion


Hach method 8190.


Ordering information: Pack of 100 powder pillows.

Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.

Formula: K₂S₂O₈
MW: 270.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011386
CAS Number: 7727-21-1
UN: 1492
ADR: 5.1,III

Order Now

Specification Test Results

Number of tests 100
Parameter Total Phosphate
Platform Powder Pillows
Quantity 100 /pk

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR