PFA Funnels, Savillex
Supplier: SAVILLEX
700-440 700-480
30617-530EA 113.94 USD
30617-530 30617-532
PFA Funnels, Savillex
Funnels Filtration Funnels Filter Funnels
Service temperature range is –200 to 260°C (–328 to 500°F).