PFA Microcentrifuge Tubes, Savillex
Supplier: Savillex
200-915-50
30617-504EA 28.08 USD
30617-504
PFA Microcentrifuge Tubes, Savillex
Tubes Microcentrifuge Tubes
PFA microcentrifuge tube fits most standard rotors.
Service temperature range is –200 to 260 °C (–328 to 500 °F).
Nonsterile. Autoclavable. Pierceable PFA snap-on closure is available, but must be ordered separately. Tube dimensions are 12.7Ø×42.3H mm (0.5×1.7").
Ordering information: Closure is not included, and must be ordered separately.