Potassium peroxodisulfate, powder pillow

Supplier: Hach
Danger

Synonyms: Dipotassium peroxodisulphateDipotassium peroxodisulfatePeroxydisulphuric acid dipotassium saltPotassium persulfatedi-Potassium peroxodisulfatePeroxydisulfuric acid dipotassium saltPotassium persulphatedi-Potassium peroxodisulphate

2084769 2084769 2084769
104085-750PK 91.05 USD
CA20847-69 104085-750 97026-350
Potassium peroxodisulfate, powder pillow
Potassium peroxodisulphate

For total phosphate determination by acid persulfate digestion and phosphonate determination by the UV Oxidation method.


Ordering information: Pack of 100 powder pillows.

Hazardous Items:
Items with this mark may be considered hazardous under some shipping conditions.
If necessary, we will change your selected shipping method to accomodate these items.

Formula: K₂S₂O₈
MW: 270.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011386
CAS Number: 7727-21-1
UN: 1492
ADR: 5.1,III

Order Now

Specification Test Results

Method Name Persulfate Oxidation
Number of tests 100
Parameter Phosphonate, Total Phosphorus
Platform Powder Pillows
Quantity 100 /pk

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR