BOD Nutrient Buffer Pillows, 300 ml, Hach
97026-674PK
36.77
USD
97026-674
BOD Nutrient Buffer Pillows, 300 ml, Hach
Environmental Test Kits
For preparation of BOD5 dilution water
Ready for immediate use. 0.5-mL size powder pillows. Each pillow prepares 300 mL of dilution water.
Ordering information: Pack of 50 pillows.