Classic Wood Wall Cabinets, Diversified Woodcrafts
Supplier: VWR International
D05-3612 D08-3612
89373-714EA 611.53 USD
89373-714 89373-746 89373-778
Classic Wood Wall Cabinets, Diversified Woodcrafts
Furniture Casework
Classic oak laboratory grade cabinets.
  • Classic neutral color
Vertical grain matched select oak veneers finished with a VOC-free, environmentally friendly chemical-resistant finish. Cabinets stained with a light neutral finish. Plywood core cabinet body with removable backs, full extension drawer slides and stainless steel hinges. 96mm classic wire pulls, twin-pin seismic rated shelf pins, and nylon roller door catches.
Certifications: SEFA Tested.
Ordering information: Units are shipped wrapped and palletized via common carrier trucking. All cabinets include a Lifetime Warranty on manufacturing parts and processes.