Kanamycin sulfate, powder

Supplier: Corning

Synonyms: Kanamycin Monosulphate(2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphateKanamycin AKanamycin Monosulfate(2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulfate

61-176-RG 61-176-RG
45000-640EA 140.07 USD
CA45000-640 45000-640
Kanamycin sulfate, powder
Kanamycin sulphate

Kanamycin is a member of the aminoglycosides a group of molecules and interacts with the 30S subunit of prokaryotic ribosomes


It induces errors in translation and thus hinders protein synthesis. The KanR-Tn5 gene product (aminoglycoside phosphotransferase) confers resistance to kanamycin. Kanamycin Sulfate is effective against Gram-negative and -positive bacteria.


Packaging: Shipping temperature: Ambient

Formula: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00070253
CAS Number: 25389-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR