Gentamicin sulfate solution 50 mg/ml in deionized water

Supplier: Corning

Synonyms: NMD Inhibitor IIGaramycinGentiomycin CGentamicin sulfate saltGentamicin sulphate salt

30-005-CR 30-005-CR
45000-636PK 665.43 USD
45000-636 CA45000-636
Gentamicin sulfate solution 50 mg/ml in deionized water
Gentamicin sulphate

Gentamicin works as an antimicrobial agent by binding to the 30S ribosomal subunit of certain bacteria.


It is formulated to contain 50 mg/ml gentamicin in deionized water and is recommended for use in cell culture applications at 1 ml/L.


This product is effective against mycoplasma, Gram-negative and Gram-positive bacteria and is stable at 37°C for 5 days.


Packaging: Shipping temperature: Ambient

Storage Temperature: Ambient
MDL Number: MFCD00270181
CAS Number: 1405-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR