Nitric acid 0.1 N, VWR Chemicals BDH®

Supplier: VWR International
Warning

Synonyms: Nitric acid solution

BDHVBDH7208-1
BDH7208-1CS 1347.83 USD
BDH7208-1
Nitric acid 0.1 N, VWR Chemicals BDH®
Nitric acid

Made with high purity acid and deionized water. Suitable as a titrant.

Formula: HNO₃
MW: 63.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

Appearance/color Clear liquid
N.I.S.T. Traceable Yes
Normality 0.0995 - 0.1005 N
Shelf-life 18 months

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR