HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99% (dry basis), powder, Ultrapure
Supplier: Thermo Fisher Scientific
Synonyms:
HEPES free acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulfonic acid, 4-(2-Hydroxyethyl)piperazin-1-ylethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid), 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
J16926-A1
J16926-22
J16926-30
J16926-A7
101170-476EA
646.75
USD
101170-476
101175-052
101170-478
101173-706
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid ≥99% (dry basis), powder, Ultrapure
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)
White crystalline powder
Formula:
C₈H₁₈N₂O₄S Melting Pt: ∼ 236 °C (dec.) Density: 1.448 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006158 CAS Number: 7365-45-9 EINECS: 230-907-9 Merck Index: 14,4654 |
|
Specification Test Results
Assay (Unspecified) | ≥99.0% (Dry Basis) |
Identification (FTIR) | Conforms |
pKa (20 °C) | 7.55 ±0.15 |
A (250 nm, 1 M, 1 cm) | ≤0.2 |
Loss on Drying | ≤0.5% |
Residue on Ignition | ≤0.1% |
Chloride (Cl) | ≤0.005% |
Phosphate (PO₄) | ≤0.001% |
Sulfate (SO₄) | ≤0.01% |
Iron (Fe) | ≤5 ppm |
Heavy Metals (Pb) | ≤10 ppm |
Assay (Unspecified) Absorbance (250 nm, 1 M, 1 cm) | ≤0.2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...