Urea 99%, Ultrapure

Supplier: Thermo Scientific Chemicals

J65769-22 J65769-36 J65769-A4
AAJ65769-22EA 58.21 USD
AAJ65769-22 AAJ65769-36 AAJ65769-A4
Urea 99%, Ultrapure
Urea

Crystalline


Purity: 99% CAS No.: 57-13-6 Molecular Formula: CH4N2O Formula Weight: 60.06 Physical Form: Crystalline Density: 1.335 Appearance: White Odor: Ammonia-like pH: 9,2-9,5 Melting Point: 132-135° Solubility: Soluble in water Beilstein Reference: 635724 Merck Reference: 14,9867 MDL No.: MFCD00008022

Formula: NH₂CONH₂
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 132…135 °C
Density: 1.335
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 14,9867
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR