D-(+)-Sucrose for molecular biology

Supplier: Thermo Scientific Chemicals

J65148-36 J65148-A1
AAJ65148-36EA 84.41 USD
AAJ65148-36 AAJ65148-A1
D-(+)-Sucrose for molecular biology

For protein purification by sucrose density gradients


Crystalline solid


For protein purification by sucrose density gradients

Formula: C₁₂H₂₂O₁₁
Melting Pt: 185…187 °C
Density: 1.58
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR