BSA blocking buffer, 5% in PBS, with 0.05% Tween-20, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J63711-AK
J63711-AP
AAJ63711-AKEA
127
USD
AAJ63711-AK
AAJ63711-AP
BSA blocking buffer, 5% in PBS, with 0.05% Tween-20, Thermo Scientific
Buffers
Electrophoresis Buffers
Blocking Buffers
Liquid
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...