Zaprinast ≥98%

Supplier: Thermo Scientific Chemicals
Warning

J63326-MA J63326-MF
AAJ63326-MAEA 68.4 USD
AAJ63326-MA AAJ63326-MF
Zaprinast ≥98%
Zaprinast

Phosphodiesterase inhibitor


Phosphodiesterase inhibitor


Phosphodiesterase inhibitor

Formula: C₁₃H₁₃N₅O₂
Storage Temperature: Ambient
MDL Number: MFCD00214073
CAS Number: 37762-06-4
EINECS: 253-655-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR