Blocking Ragents, Denhardt's Solution (50X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
J63135-AD J63135-AE
AAJ63135-ADEA 209.02 USD
AAJ63135-AD AAJ63135-AE
Blocking Ragents, Denhardt's Solution (50X), Thermo Scientific
Buffers Electrophoresis Buffers Blocking Buffers
Denhardt's solution is used to reduce non-specific binding of detection reagents to the northern and southern blot membranes.
  • Contains 1% ficolll (type 400, pharmacia), 1% polyvinylpyrrolidone, and 1% bovine serum albumin
  • Fully miscible
Denhardt's solution is a mixture of blocking agents used in membrane-based hybridization protocols.