Rifamycin SV sodium ≥98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2S,12Z,14E,16S,17S,18R,19R,20R,21S,22R,23S,24E)-21-(Acetyloxy)-5,6,9,17,19-pentahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-2,7-(epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-1,11(2H)-dione sodium salt, Rifamycin sodium
J63095-03
J63095-06
AAJ63095-03EA
105.02
USD
AAJ63095-03
AAJ63095-06
Rifamycin SV sodium ≥98%
Rifamycin SV sodium
Antibiotic effective against mycobacteria. Can also inhibit DNA polymerase
- Orange to dark red
Orange to dark red powder
Formula:
C₃₇H₄₆NNaO₁₂ MW: 719.8 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00056847 CAS Number: 14897-39-3 EINECS: 238-965-7 Merck Index: 14,8218 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...