TRIS-buffered saline (TBS, 10X) pH 8.0, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62780-K2 J62780-K3 J62780-K7
AAJ62780-K2EA 112.23 USD
AAJ62780-K2 AAJ62780-K3 AAJ62780-K7
TRIS-buffered saline (TBS, 10X) pH 8.0, Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers

Liquid


CAS No.: 77-86-1
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00004679

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR