Bacitracin

Supplier: Thermo Scientific Chemicals
Warning

J62432-06 J62432-14
AAJ62432-06EA 114.73 USD
AAJ62432-06 AAJ62432-14
Bacitracin
Bacitracin

Inhibits cell wall synthesis in Gram-positive bacteria.


White to off-white powder


Soluble in water, methanol or ethanol

Formula: C₆₆H₁₀₃N₁₇O₁₆S₁
MW: 1421.60 g/mol
Melting Pt: 222 °C
Storage Temperature: Freezer
MDL Number: MFCD00062640
CAS Number: 1405-87-4
Merck Index: 14,934

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR