Sodium chloride 5 M in aqueous solution, autoclaved
Supplier: Thermo Scientific Chemicals
Synonyms:
Halite
, Sea salt
, NaCl
, Saline
J61890-AK
J61890-AK
AAJ61890-AKEA
63.56
USD
CAAAJ61890-AK
AAJ61890-AK
Sodium chloride 5 M in aqueous solution, autoclaved
Sodium chloride
Liquid
Formula:
NaCl MW: 58.44 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...