Sodium chloride 5 M in aqueous solution, autoclaved pH 8,0

Supplier: Thermo Scientific Chemicals

Synonyms: HaliteSea saltNaClSaline

J61807-AK J61807-AK
AAJ61807-AKEA 87.27 USD
AAJ61807-AK CAAAJ61807-AK
Sodium chloride 5 M in aqueous solution, autoclaved pH 8,0
Sodium chloride

Liquid

Formula: NaCl
MW: 58.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR