Solvent black 5
Supplier: Thermo Scientific Chemicals
J61186-A1
J61186-14
J61186-22
AAJ61186-A1EA
81.9
USD
AAJ61186-A1
AAJ61186-14
AAJ61186-22
Solvent black 5
Solvent black 5
Powder
CAS No.: 11099-03-9
Physical Form: Powder
Appearance: Black
MDL No.: MFCD00149005
Formula:
C₈H₁₉ClN₂O Storage Temperature: Ambient |
MDL Number:
MFCD00071316 CAS Number: 11099-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...