TE Buffer, (20X), pH 7.4, Autoclaved, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60738-AK
J60738-AP
AAJ60738-AKEA
106.09
USD
AAJ60738-AK
AAJ60738-AP
TE Buffer, (20X), pH 7.4, Autoclaved, Thermo Scientific
Buffers
Biological Buffers
Liquid
Physical Form: Liquid
MDL No.: MFCD00236359
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...