Disuccinimidyl glutarate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Di(N-succinimidyl) glutarate, Bis(2,5-dioxopyrrolidin-1-yl) glutarate, DSG Crosslinker, DSG

H58208-MC H58208-ME
AAH58208-MCEA 188.61 USD
AAH58208-MC AAH58208-ME
Disuccinimidyl glutarate 97%
DSG (Disuccinimidyl glutarate)

Purity: 97% CAS No.: 79642-50-5 Molecular Formula: C13H14N2O8 Formula Weight: 326.26 Storage Temperature: -30°C to -10°C Beilstein Reference: 8575145 MDL No.: MFCD00153597

Formula: C₁₃H₁₄N₂O₈
MW: 326.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00153597
CAS Number: 79642-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR