5-Methylphenazinium methyl sulphate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenazine methosulfate, PMS, 5-Methylphenazin-5-ium methyl sulfate, 5-Methylphenazin-5-ium methyl sulphate, Phenazine methosulphate, Phenazine methylsulfate, Phenazine methylsulphate

H56718-06 H56718-14
AAH56718-06EA 141 USD
AAH56718-06 AAH56718-14
5-Methylphenazinium methyl sulphate 98+%
5-Methylphenazinium methyl sulphate

Purity: 98+% CAS No.: 299-11-6 Molecular Formula: C13H11N2.CH3SO4 Formula Weight: 306.34 Storage Temperature: -30°C to -10°C

Formula: C₁₄H₁₄N₂O₄S
Melting Pt: 198 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011923
CAS Number: 299-11-6
EINECS: 206-072-1
Merck Index: 12,06185

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR