1-Acetyladamantane 99%
Supplier: Thermo Scientific Chemicals
H55889-03
H55889-06
AAH55889-03EA
33.6
USD
AAH55889-03
AAH55889-06
AAH55889-14
1-Acetyladamantane 99%
1-Acetyladamantane
Purity: 99%
CAS No.: 1660-04-4
Molecular Formula: C12H18O
Formula Weight: 178.27
Flash Point: 108°(226°F)
Melting Point: 53-55°
MDL No.: MFCD00074739
Formula:
C₁₂H₁₈O Melting Pt: 53…55 °C Flash Pt: 108 °C(226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00074739 CAS Number: 1660-04-4 EINECS: 216-761-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...