2-Amino-4-chloropyrimidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-chloropyrimidin-2-amine
H55007-03
H55007-06
AAH55007-03EA
70.3
USD
AAH55007-03
AAH55007-06
AAH55007-MD
2-Amino-4-chloropyrimidine 98%
2-Amino-4-chloropyrimidine
Purity: 98%
CAS No.: 3993-78-0
Molecular Formula: C4H4ClN3
Formula Weight: 129.55
Storage Temperature: 2°C to 8°C
Melting Point: 155-160° dec.
MDL No.: MFCD00038021
Formula:
C₄H₄ClN₃ Melting Pt: 155…160 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00038021 CAS Number: 3993-78-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...