2-Pyrimidinecarboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pyrimidine-2-carboxylic acid, 2-Carboxypyrimidine
H54745-03
H54745-06
AAH54745-03EA
65.4
USD
AAH54745-03
AAH54745-06
AAH54745-MD
2-Pyrimidinecarboxylic acid 97%
2-Pyrimidinecarboxylic acid
Purity: 97%
CAS No.: 31519-62-7
Molecular Formula: C5H4N2O2
Formula Weight: 124.10
MDL No.: MFCD00856161
Formula:
C₅H₄N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00856161 CAS Number: 31519-62-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...