1,1-Dimethylpropargylamine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methylbut-3-yn-2-amine
H53448-06
H53448-14
AAH53448-06EA
69.3
USD
AAH53448-06
AAH53448-14
1,1-Dimethylpropargylamine 95%
1,1-Dimethylpropargylamine
Purity: 95%
CAS No.: 2978-58-7
Molecular Formula: C5H9N
Formula Weight: 83.13
Flash Point: 2°(36°F)
Refractive Index: 1.4237
Boiling Point: 79-80°
UN No.: 2733
MDL No.: MFCD00008052
Formula:
C₅H₉N MW: 83.134 g/mol Boiling Pt: 79…80 °C Density: 0.79 Flash Pt: 2 °C(36 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008052 CAS Number: 2978-58-7 EINECS: 221-029-7 UN: 2733 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...